alpha-D(+)-Glucose, 99+%, anhydrous
alpha-D(+)-Glucose, 99+%, anhydrous
Thermo Scientific Chemicals

alpha-D(+)-Glucose, 99+%, anhydrous

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
25 kg
Catalog number 170080010
Price (USD)/ Each
37.65
Online exclusive
41.80 
Save 4.15 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
37.65
Online exclusive
41.80 
Save 4.15 (10%)
Add to cart
alpha-D(+)-Glucose, 99+%, anhydrous
Catalog number170080010
Price (USD)/ Each
37.65
Online exclusive
41.80 
Save 4.15 (10%)
-
Add to cart
Chemical Identifiers
CAS492-62-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Specific optical rotationon anhydrous substance
Specific optical rotation+52.5° to +53.3° (20°C, 589 nm) (c=10, H2O/diluteNH3)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only