Thermo Scientific Chemicals

Methyl p-toluenesulfonate, 97%, Thermo Scientific Chemicals

Catalog number: 170101000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl p-toluenesulfonate, 97%, Thermo Scientific Chemicals

Catalog number: 170101000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 170101000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
80-48-8
IUPAC Name
methyl 4-methylbenzene-1-sulfonate
Molecular Formula
C8H10O3S
InChI Key
VUQUOGPMUUJORT-UHFFFAOYSA-N
SMILES
COS(=O)(=O)C1=CC=C(C)C=C1
Infrared spectrum
Conforms
Melting point
24°C to 30°C
Titration excess NaOH
96.0 to 104.0 %
GC
>=96.0 %
Water
=<0.5 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text