β-D-Ribofuranose 1-acetate 2,3,5-tribenzoate, 99%
β-D-Ribofuranose 1-acetate 2,3,5-tribenzoate, 99%
Thermo Scientific Chemicals

β-D-Ribofuranose 1-acetate 2,3,5-tribenzoate, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
Packaging:
Glass bottle
Catalog number 170110250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6974-32-9
IUPAC Name[5-(acetyloxy)-3,4-bis(benzoyloxy)oxolan-2-yl]methyl benzoate
Molecular FormulaC28H24O9
InChI KeyGCZABPLTDYVJMP-UHFFFAOYNA-N
SMILESCC(=O)OC1OC(COC(=O)C2=CC=CC=C2)C(OC(=O)C2=CC=CC=C2)C1OC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
HPLC>=98.5 %
Water=<1 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only