Acetohydroxamic acid, 98+%
Acetohydroxamic acid, 98+%
Acetohydroxamic acid, 98+%
Acetohydroxamic acid, 98+%
Thermo Scientific Chemicals

Acetohydroxamic acid, 98+%

CAS: 546-88-3 | C2H5NO2 | 75.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 170130250
Price (USD)
220.65
Online Exclusive
245.00
Save 24.35 (10%)
Each
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
220.65
Online Exclusive
245.00
Save 24.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS546-88-3
IUPAC NameN-hydroxyacetamide
Molecular FormulaC2H5NO2
InChI KeyRRUDCFGSUDOHDG-UHFFFAOYSA-N
SMILESCC(=O)NO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
Melting point85°C to 90°C
Titration with NaOH98.0 to 102.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link