Thermo Scientific Chemicals

tert-Butyl peroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: 170140010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

tert-Butyl peroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: 170140010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 170140010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
614-45-9
IUPAC Name
tert-butyl benzenecarboperoxoate
Molecular Formula
C11H14O3
InChI Key
GJBRNHKUVLOCEB-UHFFFAOYSA-N
SMILES
CC(C)(C)OOC(=O)C1=CC=CC=C1
Appearance (Color)
Clear yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Additional info
saponifiable chlorine: < 0.035%
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiators
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text