Acetamidine hydrochloride, 98+%, Thermo Scientific Chemicals
Acetamidine hydrochloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetamidine hydrochloride, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
500 g
Packaging:
Glass bottle
Catalog number 170211000
Price (USD)/ Each
59.90
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
59.90
Add to cart
Acetamidine hydrochloride, 98+%, Thermo Scientific Chemicals
Catalog number170211000
Price (USD)/ Each
59.90
-
Add to cart
Chemical Identifiers
CAS124-42-5
IUPAC Namehydrogen ethanimidamide chloride
Molecular FormulaC2H7ClN2
InChI KeyWCQOBLXWLRDEQA-UHFFFAOYSA-N
SMILES[H+].[Cl-].CC(N)=N
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric98.0 to 102.0 %
Appearance (Color)White to cream
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only