Thermo Scientific Chemicals

Acetamidine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: 170215000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Acetamidine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: 170215000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 170215000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
124-42-5
IUPAC Name
hydrogen ethanimidamide chloride
Molecular Formula
C2H7ClN2
InChI Key
WCQOBLXWLRDEQA-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CC(N)=N
Appearance (Color)
White to cream
Appearance (Form)
Crystalline powder or crystals and/or chunks
Infrared spectrum
Conforms
Titration Argentometric
98.0 to 102.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text