Diethyl pyrocarbonate, 97%, pure
This product ships via ground from US northeast.
Diethyl pyrocarbonate, 97%, pure
Diethyl pyrocarbonate, 97%, pure
Diethyl pyrocarbonate, 97%, pure
Thermo Scientific Chemicals

Diethyl pyrocarbonate, 97%, pure

CAS: 1609-47-8 | C6H10O5 | 162.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 170250250
Price (USD)
90.65
Special offer
Online exclusive
Ends: 26-Sep-2025
111.00
Save 20.35 (18%)
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
90.65
Special offer
Online exclusive
Ends: 26-Sep-2025
111.00
Save 20.35 (18%)
Each
Add to cart
Chemical Identifiers
CAS1609-47-8
IUPAC Namediethyl dicarbonate
Molecular FormulaC6H10O5
InChI KeyFFYPMLJYZAEMQB-UHFFFAOYSA-N
SMILESCCOC(=O)OC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to amber
Appearance (Form)Liquid
Infrared spectrumConforms
Titration excess NaOH>=96.0 %
Refractive index1.3960 to 1.4000 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link