Thermo Scientific Chemicals

Hydroxylamine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 170360050
5 kg, Each
Thermo Scientific Chemicals

Hydroxylamine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 170360050
5 kg, Each
Quantity
Catalog number: 170360050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5470-11-1
IUPAC Name
hydroxylamine hydrochloride
Molecular Formula
ClH4NO
InChI Key
WTDHULULXKLSOZ-UHFFFAOYSA-N
SMILES
Cl.NO
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder or crystals and/or chunks
Infrared spectrum
Conforms
Melting point
151°C to 157°C (Dec)
Titration with NaOH
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text