Thermo Scientific Chemicals

Methyl 3,5-dihydroxybenzoate, 97%, Thermo Scientific Chemicals

Catalog number: 170440250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 3,5-dihydroxybenzoate, 97%, Thermo Scientific Chemicals

Catalog number: 170440250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 170440250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2150-44-9
IUPAC Name
methyl 3,5-dihydroxybenzoate
Molecular Formula
C8H8O4
InChI Key
RNVFYQUEEMZKLR-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(O)=CC(O)=C1
Appearance (Color)
Light yellow to beige
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
164°C to 169°C
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text