Methyl 3,5-dihydroxybenzoate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl 3,5-dihydroxybenzoate, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 170441000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2150-44-9
IUPAC Namemethyl 3,5-dihydroxybenzoate
Molecular FormulaC8H8O4
InChI KeyRNVFYQUEEMZKLR-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(O)=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Color)Light yellow to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point164°C to 169°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only