Thermo Scientific Chemicals

2,4-Dimethoxybenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: 170461000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2,4-Dimethoxybenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: 170461000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 170461000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7314-44-5
IUPAC Name
(2,4-dimethoxyphenyl)methanol
Molecular Formula
C9H12O3
InChI Key
RNKOUSCCPHSCFE-UHFFFAOYSA-N
SMILES
COC1=CC=C(CO)C(OC)=C1
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid or low melting solid
Infrared spectrum
Conforms
Melting point
34°C to 38°C
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text