1-(4-Chlorophenyl)-1-cyclopropanecarboxylic acid, 99%, Thermo Scientific Chemicals
1-(4-Chlorophenyl)-1-cyclopropanecarboxylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(4-Chlorophenyl)-1-cyclopropanecarboxylic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 170690500
Price (USD)/ Each
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS72934-37-3
IUPAC Name1-(4-chlorophenyl)cyclopropane-1-carboxylic acid
Molecular FormulaC10H9ClO2
InChI KeyYAHLWSGIQJATGG-UHFFFAOYSA-N
SMILESOC(=O)C1(CC1)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point152°C to 157°C
Titration with NaOH>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only