1-(4-Methylphenyl)-1-cyclopropanecarboxylic acid, 98%, Thermo Scientific Chemicals
1-(4-Methylphenyl)-1-cyclopropanecarboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(4-Methylphenyl)-1-cyclopropanecarboxylic acid, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
10 g
Packaging:
Glass bottle
Catalog number 170700100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name1-(4-methylphenyl)cyclopropane-1-carboxylic acid
Molecular FormulaC11H12O2
InChI KeyAYUGAOYMYXSOKU-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C1(CC1)C(O)=O
Molecular Weight (g/mol)176.22
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point109°C to 114°C
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only