Soybean Trypsin Inhibitor, powder
Save up to 29% on your order of cell culture products. View promotion details >
Product Image
Gibco™

Soybean Trypsin Inhibitor, powder

Soybean Trypsin Inhibitor is one of several protease inhibitors found in soybeans. Trypsin is inhibited at a molar ratio ofRead more
Have Questions?
Catalog NumberQuantity
170750291 g
Catalog number 17075029
Price (USD)
223.00
Each
-
Add to cart
Quantity:
1 g
Price (USD)
223.00
Each
Add to cart
Ask our AI about this Product
Soybean Trypsin Inhibitor is one of several protease inhibitors found in soybeans. Trypsin is inhibited at a molar ratio of 1:1, while chymotrypsin and plasmin are inhibited to a lesser extent. Soybean trypsin inhibitor will also inhibit other serine proteases.

Gibco™ Soybean Trypsin Inhibitor, powder has an activity greater than 7,000 BAEE units per mg. A 0.5 mg/mL Soybean Trypsin Inhibitor stock solution should be made by dissolving the non-sterile powder in 0.01M Borate Buffer or Dulbecco's Phosphate Buffered Saline (DPBS), without calcium and magnesium. Filter sterilize the stock solution and store for up to a week at 2 - 8°C.

Product Use
For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells
Quantity1 g
Shipping ConditionDry Ice
FormFrozen, powder
Product TypeTrypsin inhibitor
SterilityNon-sterile
Without AdditivesNo Calcium, No Magnesium
Unit SizeEach
Contents & Storage
Storage conditions: -5 to -30°C
Shipping conditions: Frozen
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3184223Certificate of AnalysisJun 21, 202517075029
3124578Certificate of AnalysisMar 30, 202517075029
2789744Certificate of AnalysisJan 29, 202517075029
2810678Certificate of AnalysisJan 28, 202517075029
2836589Certificate of AnalysisJan 27, 202517075029
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

This product is derived from a Kunitz purification, but in powder form

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Please use this selection chart that compares our cell dissociation reagents (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/reagents/trypsin.html).

Find additional tips, troubleshooting help, and resources within ourMammalian Cell Culture Basics Support Center.

Share catalog number, name or link

1x1 image pixel for data collection