Thermo Scientific Chemicals

4-Nitrophenyl chloroformate, 97%, Thermo Scientific Chemicals

Catalog number: 170800050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Nitrophenyl chloroformate, 97%, Thermo Scientific Chemicals

Catalog number: 170800050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 170800050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7693-46-1
IUPAC Name
4-nitrophenyl carbonochloridate
Molecular Formula
C7H4ClNO4
InChI Key
NXLNNXIXOYSCMB-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(OC(Cl)=O)C=C1
Appearance (Color)
White to yellow to light beige
Appearance (Form)
Crystalline powder or crystals and/or chunks
Infrared spectrum
Conforms
Melting point
77°C to 82°C
Titration Argentometric
>=96.0 % (ex Cl)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text