2-Bromothiazole, 98+%, Thermo Scientific Chemicals
2-Bromothiazole, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromothiazole, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 170910050
Price (USD)/ Each
42.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
49.70 
Save 7.05 (14%)
-
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
42.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
49.70 
Save 7.05 (14%)
Add to cart
2-Bromothiazole, 98+%, Thermo Scientific Chemicals
Catalog number170910050
Price (USD)/ Each
42.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
49.70 
Save 7.05 (14%)
-
Add to cart
Chemical Identifiers
CAS3034-53-5
IUPAC Name2-bromo-1,3-thiazole
Molecular FormulaC3H2BrNS
InChI KeyRXNZFHIEDZEUQM-UHFFFAOYSA-N
SMILESBrC1=NC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Appearance (Color)Clear colorless to orange to light brown
GC>=98.0 %
Refractive index1.5915 to 1.5945 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only