Thermo Scientific Chemicals

9-Fluorenylmethyl chloroformate, 98%, Thermo Scientific Chemicals

Catalog number: 170940050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

9-Fluorenylmethyl chloroformate, 98%, Thermo Scientific Chemicals

Catalog number: 170940050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 170940050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
28920-43-6
IUPAC Name
(9H-fluoren-9-yl)methyl carbonochloridate
Molecular Formula
C15H11ClO2
InChI Key
IRXSLJNXXZKURP-UHFFFAOYSA-N
SMILES
ClC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Appearance (Color)
White to light yellow
Melting point
60°C to 64°C
HPLC
>=97.5 %
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text