Thermo Scientific Chemicals

Iodoacetic acid, sodium salt, 99%, Thermo Scientific Chemicals

Catalog number: 170970250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Iodoacetic acid, sodium salt, 99%, Thermo Scientific Chemicals

Catalog number: 170970250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 170970250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
305-53-3
IUPAC Name
sodium 2-iodoacetate
Molecular Formula
C2H2INaO2
InChI Key
AGDSCTQQXMDDCV-UHFFFAOYSA-M
SMILES
[Na+].[O-]C(=O)CI
Appearance (Color)
Light yellow to yellow-brown
Infrared spectrum
Conforms
Titration with HClO4
>=98.5 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text