Thermo Scientific Chemicals

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid, disodium salt hydrat, Thermo Scientific Chemicals

Catalog number: 171010050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid, disodium salt hydrat, Thermo Scientific Chemicals

Catalog number: 171010050
5 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 171010050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1264198-47-1
IUPAC Name
disodium 4-[3-(pyridin-2-yl)-5-(4-sulfonatophenyl)-1,2,4-triazin-6-yl]benzene-1-sulfonate
Molecular Formula
C20H12N4Na2O6S2
InChI Key
DDIHLFCSEHAOJZ-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=C(N=C(N=N1)C1=CC=CC=N1)C1=CC=C(C=C1)S([O-])(=O)=O
Suitability
(for Cu + Fe determination) Passes test
Appearance (Color)
Yellow to yellow-green
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration with HClO4
>=97.0 % (On dried substance)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text