Thermo Scientific Chemicals

Quinaldic acid, 99%, Thermo Scientific Chemicals

Catalog number: 171050100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Quinaldic acid, 99%, Thermo Scientific Chemicals

Catalog number: 171050100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 171050100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
93-10-7
IUPAC Name
quinoline-2-carboxylic acid
Molecular Formula
C10H7NO2
InChI Key
LOAUVZALPPNFOQ-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C2C=CC=CC2=N1
Appearance (Color)
Light brown
Appearance (Form)
Crystalline powder or needles
Infrared spectrum
Conforms
Melting point
157°C to 159°C
Titration with NaOH
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text