1,2-Naphthoquinone, 95%, Tech., Thermo Scientific Chemicals
1,2-Naphthoquinone, 95%, Tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2-Naphthoquinone, 95%, Tech., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
10 g
25 g
Packaging:
Glass bottle
Catalog number 171230010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS524-42-5
IUPAC Name1,2-dihydronaphthalene-1,2-dione
Molecular FormulaC10H6O2
InChI KeyKETQAJRQOHHATG-UHFFFAOYSA-N
SMILESO=C1C=CC2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Appearance (Form)Powder
GC>=94 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only