Thermo Scientific Chemicals

4-Biphenylcarbonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 171250500
50 g, Each
Thermo Scientific Chemicals

4-Biphenylcarbonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 171250500
50 g, Each
Quantity
Catalog number: 171250500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14002-51-8
IUPAC Name
[1,1'-biphenyl]-4-carbonyl chloride
Molecular Formula
C13H9ClO
InChI Key
JPVUWCPKMYXOKW-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC=C(C=C1)C1=CC=CC=C1
Appearance (Color)
White to yellow
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
110°C to 114°C
Titration Argentometric
>=97.5 % (ex Cl)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text