Thermo Scientific Chemicals

6-chloropurine, 99+%, Thermo Scientific Chemicals

Catalog number: 171280050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

6-chloropurine, 99+%, Thermo Scientific Chemicals

Catalog number: 171280050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 171280050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
87-42-3
IUPAC Name
6-chloro-7H-purine
Molecular Formula
C5H3ClN4
InChI Key
ZKBQDFAWXLTYKS-UHFFFAOYSA-N
SMILES
ClC1=C2NC=NC2=NC=N1
Heavy metals (as Pb)
=<10 ppm
Appearance (Color)
Yellow to orange
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with HClO4
99.0 to 101.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text