Thermo Scientific Chemicals

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: 171440500
50 g, Glass Bottle, Each
Thermo Scientific Chemicals

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: 171440500
50 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 171440500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25952-53-8
IUPAC Name
({[3-(dimethylamino)propyl]imino}methylidene)(ethyl)amine hydrochloride
Molecular Formula
C8H18ClN3
InChI Key
FPQQSJJWHUJYPU-UHFFFAOYSA-N
SMILES
Cl.CCN=C=NCCCN(C)C
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
111°C to 116°C
Titration Argentometric
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text