Thermo Scientific Chemicals

Cyclohexanepropionic acid, 99%, Thermo Scientific Chemicals

Catalog number: 171452500
250 g, Each
Thermo Scientific Chemicals

Cyclohexanepropionic acid, 99%, Thermo Scientific Chemicals

Catalog number: 171452500
250 g, Each
Quantity
Catalog number: 171452500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
701-97-3
IUPAC Name
3-cyclohexylpropanoic acid
Molecular Formula
C9H16O2
InChI Key
HJZLEGIHUQOJBA-UHFFFAOYSA-N
SMILES
OC(=O)CCC1CCCCC1
Refractive index
1.4630 to 1.4650 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Appearance
May darken on storage
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text