Appearance (Form)Crystals and/or chunks or fused solid
Infrared spectrumConforms
Melting point40°C to 46°C
Titration with HClO496.0 to 104.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.