10-Undecenoyl chloride, 98%, Thermo Scientific Chemicals
10-Undecenoyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

10-Undecenoyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number 171585000
Price (USD)/ Each
366.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
431.00 
Save 64.35 (15%)
-
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
366.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
431.00 
Save 64.35 (15%)
Add to cart
10-Undecenoyl chloride, 98%, Thermo Scientific Chemicals
Catalog number171585000
Price (USD)/ Each
366.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
431.00 
Save 64.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameundec-10-enoyl chloride
Molecular FormulaC11H19ClO
InChI KeyMZFGYVZYLMNXGL-UHFFFAOYSA-N
SMILESClC(=O)CCCCCCCCC=C
Molecular Weight (g/mol)202.72
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4530 to 1.4560 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only