Thermo Scientific Chemicals

Bromomethyl methyl ether, 90%, Thermo Scientific Chemicals

Catalog number: 171650250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Bromomethyl methyl ether, 90%, Thermo Scientific Chemicals

Catalog number: 171650250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 171650250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13057-17-5
IUPAC Name
bromo(methoxy)methane
Molecular Formula
C2H5BrO
InChI Key
JAMFGQBENKSWOF-UHFFFAOYSA-N
SMILES
COCBr
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.4550 to 1.4570 (20°C, 589 nm)
GC
>=88 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text