Thermo Scientific Chemicals

Dithioerythritol, 99+%, Thermo Scientific Chemicals

Catalog number: 171660050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Dithioerythritol, 99+%, Thermo Scientific Chemicals

Catalog number: 171660050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 171660050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6892-68-8
IUPAC Name
(2R,3S)-1,4-disulfanylbutane-2,3-diol
Molecular Formula
C4H10O2S2
InChI Key
VHJLVAABSRFDPM-ZXZARUISSA-N
SMILES
O[C@@H](CS)[C@H](O)CS
Infrared spectrum
Conforms
Melting point
82°C to 86°C
Titration Iodimetric
>=99.0 %
Solubility
(1 % in water) Clear to slightly hazy colorless
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text