Dimethylglyoxime, 99+%, Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethylglyoxime, 99+%, Thermo Scientific Chemicals

Dimethylglyoxime, 99+%, C4H8N2O2, CAS Number-95-45-4, 2,3-butanedione dioxime, chugaev's reagent, biacetyl, dioxime, 2,3-diisonitrosobutane, diacetyldioxime, nsc 9, diacetyl dioxime, unii-2971mft1ky, biacetyl dioxime, dimethylglyoxime, 500g, 01,772, White, 202-420-1, 116.12, GHS Signal Word: Danger
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number 172000025
Price (USD)/ Each
444.65
Online exclusive
494.00 
Save 49.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
444.65
Online exclusive
494.00 
Save 49.35 (10%)
Add to cart
Dimethylglyoxime, 99+%, Thermo Scientific Chemicals
Catalog number172000025
Price (USD)/ Each
444.65
Online exclusive
494.00 
Save 49.35 (10%)
-
Add to cart
Chemical Identifiers
CAS95-45-4
IUPAC Name(E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine
Molecular FormulaC4H8N2O2
InChI KeyJGUQDUKBUKFFRO-GGWOSOGESA-N
SMILESC\C(=N/O)\C(\C)=N\O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point239°C to 242°C (Dec)
Gravimetry>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only