D-Phenylalanine, 99+%
D-Phenylalanine, 99+%
Thermo Scientific Chemicals

D-Phenylalanine, 99+%

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass Bottle
Catalog number 172041000
Price (USD)/ Each
242.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
285.00 
Save 42.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
242.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
285.00 
Save 42.35 (15%)
Add to cart
D-Phenylalanine, 99+%
Catalog number172041000
Price (USD)/ Each
242.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
285.00 
Save 42.35 (15%)
-
Add to cart
Chemical Identifiers
CAS673-06-3
IUPAC Name(2R)-2-amino-3-phenylpropanoic acid
Molecular FormulaC9H11NO2
InChI KeyCOLNVLDHVKWLRT-MRVPVSSYSA-N
SMILESN[C@H](CC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Loss on drying=<0.5 %
Sulfated ash=<0.1 %
Heavy metals (as Pb)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only