Thermo Scientific Chemicals

N-Carbobenzyloxy-L-aspartic acid, 98%

Catalog number: 172051000
100 g, Each
Thermo Scientific Chemicals

N-Carbobenzyloxy-L-aspartic acid, 98%

Catalog number: 172051000
100 g, Each
Quantity
Catalog number: 172051000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1152-61-0
IUPAC Name
2-{[(benzyloxy)carbonyl]amino}butanedioic acid
Molecular Formula
C12H13NO6
InChI Key
XYXYXSKSTZAEJW-UHFFFAOYNA-N
SMILES
OC(=O)CC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Appearance (Color)
White
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration with NaOH
97.5 to 102.5 %
Specific optical rotation
+8° to +10° (20°C, 589 nm) (c=1, acetic acid)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text