Thermo Scientific Chemicals

DL-2-Aminobutyric acid, 99%, Thermo Scientific Chemicals

Catalog number: 172090250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

DL-2-Aminobutyric acid, 99%, Thermo Scientific Chemicals

Catalog number: 172090250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 172090250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2835-81-6
IUPAC Name
2-aminobutanoic acid
Molecular Formula
C4H9NO2
InChI Key
QWCKQJZIFLGMSD-UHFFFAOYNA-N
SMILES
CCC(N)C(O)=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with HClO4
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text