Thermo Scientific Chemicals

DL-Valine, +99%

Catalog number: 172102500
250 g, Plastic Bottle, Each
Thermo Scientific Chemicals

DL-Valine, +99%

Catalog number: 172102500
250 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 172102500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
516-06-3
IUPAC Name
2-amino-3-methylbutanoic acid
Molecular Formula
C5H11NO2
InChI Key
KZSNJWFQEVHDMF-UHFFFAOYSA-N
SMILES
CC(C)C(N)C(O)=O
Appearance (Form)
Crystalline powder
Titration with HClO4
98.0 to 101.0 %
Heavy metals (as Pb)
=<10 ppm
UV transmittance
>=95 % (c=10, 2 N HCl, 430 nm) 1 cm cell
Loss on drying
=<0.2 % (105°C, 3 hrs)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistry
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text