Thermo Scientific Chemicals

2-Picolyl chloride hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 172120050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Picolyl chloride hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 172120050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 172120050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6959-47-3
IUPAC Name
hydrogen 2-(chloromethyl)pyridine chloride
Molecular Formula
C6H7Cl2N
InChI Key
JPMRGPPMXHGKRO-UHFFFAOYSA-N
SMILES
[H+].[Cl-].ClCC1=CC=CC=N1
Appearance (Form)
Crystalline mass
Melting point
119°C to 125°C
Infrared spectrum
Conforms
Appearance (Color)
Yellow-green to light brown
Titration with HClO4
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text