2-(2-Chloroethoxy)ethanol, 99%, Thermo Scientific Chemicals
2-(2-Chloroethoxy)ethanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(2-Chloroethoxy)ethanol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
50 mL
Packaging:
Glass bottle
Catalog number 172262500
Price (USD)/ Each
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-(2-chloroethoxy)ethan-1-ol
Molecular FormulaC4H9ClO2
InChI KeyLECMBPWEOVZHKN-UHFFFAOYSA-N
SMILESOCCOCCCl
Molecular Weight (g/mol)124.56
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<1 % (K.F.)
Color scale=<20 APHA
GC>=98.5 %
Refractive index1.4519 to 1.4539 (20°C, 589 nm)
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only