n-Butylboronic acid, 98%, Thermo Scientific Chemicals
n-Butylboronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Butylboronic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 172380050
Price (USD)/ Each
90.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
106.00 
Save 15.35 (14%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
90.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
106.00 
Save 15.35 (14%)
Add to cart
n-Butylboronic acid, 98%, Thermo Scientific Chemicals
Catalog number172380050
Price (USD)/ Each
90.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
106.00 
Save 15.35 (14%)
-
Add to cart
Chemical Identifiers
CAS4426-47-5
IUPAC Namebutylboronic acid
Molecular FormulaC4H11BO2
InChI KeyQPKFVRWIISEVCW-UHFFFAOYSA-N
SMILESCCCCB(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Glistening powder or flakes
Titration with NaOH>=97.5 %
Infrared spectrumConforms
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagent
RUO – Research Use Only