Thermo Scientific Chemicals

n-Butylboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: 172380250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

n-Butylboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: 172380250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 172380250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4426-47-5
IUPAC Name
butylboronic acid
Molecular Formula
C4H11BO2
InChI Key
QPKFVRWIISEVCW-UHFFFAOYSA-N
SMILES
CCCCB(O)O
Appearance (Form)
Glistening powder or flakes
Titration with NaOH
>=97.5 %
Infrared spectrum
Conforms
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text