7-Methoxy-1-tetralone, 97%, Thermo Scientific Chemicals
7-Methoxy-1-tetralone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

7-Methoxy-1-tetralone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 172430250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name7-methoxy-1,2,3,4-tetrahydronaphthalen-1-one
Molecular FormulaC11H12O2
InChI KeyGABLTKRIYDNDIN-UHFFFAOYSA-N
SMILESCOC1=CC=C2CCCC(=O)C2=C1
Molecular Weight (g/mol)176.22
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
GC>=96.0 %
Melting point60°C to 64°C
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only