Thermo Scientific Chemicals

4-(4-Methoxyphenyl)butyric acid, 98%, Thermo Scientific Chemicals

Catalog number: 172440050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-(4-Methoxyphenyl)butyric acid, 98%, Thermo Scientific Chemicals

Catalog number: 172440050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 172440050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4521-28-2
IUPAC Name
4-(4-methoxyphenyl)butanoic acid
Molecular Formula
C11H14O3
InChI Key
LZHMNCJMXQKSBY-UHFFFAOYSA-N
SMILES
COC1=CC=C(CCCC(O)=O)C=C1
Appearance (Form)
Powder
Titration with TBAH
>=97.5 %
Appearance (Color)
Cream to light yellow
Melting point
56°C to 61°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text