3-Furoic acid, 99%, Thermo Scientific Chemicals
3-Furoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Furoic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 172450250
Price (USD)/ Each
260.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
307.00 
Save 46.35 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
260.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
307.00 
Save 46.35 (15%)
Add to cart
3-Furoic acid, 99%, Thermo Scientific Chemicals
Catalog number172450250
Price (USD)/ Each
260.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
307.00 
Save 46.35 (15%)
-
Add to cart
Chemical Identifiers
CAS488-93-7
IUPAC Namefuran-3-carboxylic acid
Molecular FormulaC5H4O3
InChI KeyIHCCAYCGZOLTEU-UHFFFAOYSA-N
SMILESOC(=O)C1=COC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige
Infrared spectrumConforms
Melting point121°C to 124°C
Titration with NaOH>=98.5 %
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only