Thermo Scientific Chemicals

(+)-Dimethyl L-tartrate, 99%, ACROS Organics™

Catalog number: 172490250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

(+)-Dimethyl L-tartrate, 99%, ACROS Organics™

Catalog number: 172490250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 172490250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
608-68-4
IUPAC Name
1,4-dimethyl (2R,3S)-2,3-dihydroxybutanedioate
Molecular Formula
C6H10O6
InChI Key
PVRATXCXJDHJJN-ZXZARUISSA-N
SMILES
COC(=O)[C@@H](O)[C@@H](O)C(=O)OC
Appearance (Form)
Crystalline powder or crystals and/or chunks
Infrared spectrum
Conforms
Specific optical rotation
+20° to +23° (20°C, 589 nm) (c=10, water)
Appearance (Color)
White to almost white
GC
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text