Thermo Scientific Chemicals

HEPES, 99%, for biochemistry

Catalog number: 172571000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

HEPES, 99%, for biochemistry

Catalog number: 172571000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 172571000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7365-45-9
IUPAC Name
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
Molecular Formula
C8H18N2O4S
InChI Key
JKMHFZQWWAIEOD-UHFFFAOYSA-N
SMILES
OCCN1CCN(CCS(O)(=O)=O)CC1
UV
at 260 nm A: =<0.08
UV
at 280 nm A: =<0.05
Solubility
(5 % in water) Clear colorless
Appearance (Color)
White
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text