HEPES, 99%, for biochemistry
HEPES, 99%, for biochemistry
Thermo Scientific Chemicals

HEPES, 99%, for biochemistry

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
100 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 172572500
Price (USD)/ Each
238.65
Online exclusive
265.00 
Save 26.35 (10%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
238.65
Online exclusive
265.00 
Save 26.35 (10%)
Add to cart
HEPES, 99%, for biochemistry
Catalog number172572500
Price (USD)/ Each
238.65
Online exclusive
265.00 
Save 26.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7365-45-9
IUPAC Name2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
Molecular FormulaC8H18N2O4S
InChI KeyJKMHFZQWWAIEOD-UHFFFAOYSA-N
SMILESOCCN1CCN(CCS(O)(=O)=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 260 nm A: =<0.08
UVat 280 nm A: =<0.05
Solubility(5 % in water) Clear colorless
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only