Thermo Scientific Chemicals

BES, 99+%, for biochemistry

Catalog number: 172581000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

BES, 99+%, for biochemistry

Catalog number: 172581000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 172581000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10191-18-1
IUPAC Name
2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonic acid
Molecular Formula
C6H15NO5S
InChI Key
AJTVSSFTXWNIRG-UHFFFAOYSA-N
SMILES
OCCN(CCO)CCS(O)(=O)=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with CHA
>=99.0 %
Loss on drying
=<0.5 % (100°C, 2 hrs)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text