BES, 99+%, for biochemistry
BES, 99+%, for biochemistry
Thermo Scientific Chemicals

BES, 99+%, for biochemistry

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number 172585000
Price (USD)/ Each
324.65
Online exclusive
361.00 
Save 36.35 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
324.65
Online exclusive
361.00 
Save 36.35 (10%)
Add to cart
BES, 99+%, for biochemistry
Catalog number172585000
Price (USD)/ Each
324.65
Online exclusive
361.00 
Save 36.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10191-18-1
IUPAC Name2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonic acid
Molecular FormulaC6H15NO5S
InChI KeyAJTVSSFTXWNIRG-UHFFFAOYSA-N
SMILESOCCN(CCO)CCS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with CHA>=99.0 %
Loss on drying=<0.5 % (100°C, 2 hrs)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only