MES hydrate, 99+% for biochemistry
This product ships via ground from US northeast.
Save 25% on this product with promo code P5488313
MES hydrate, 99+% for biochemistry
Thermo Scientific Chemicals

MES hydrate, 99+% for biochemistry

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 172595000
Price (USD)/ Each
504.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
593.00 
Save 88.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
504.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
593.00 
Save 88.35 (15%)
Add to cart
MES hydrate, 99+% for biochemistry
Catalog number172595000
Price (USD)/ Each
504.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
593.00 
Save 88.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-(morpholin-4-yl)ethane-1-sulfonic acid
Molecular FormulaC6H13NO4S
InChI KeySXGZJKUKBWWHRA-UHFFFAOYSA-N
SMILESOS(=O)(=O)CCN1CCOCC1
Molecular Weight (g/mol)195.23
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with NaOH>=99.0 % (On dry substance)
UVat 260 nm A: =<0.05
Appearance (Form)Crystalline powder
Water=<9.0 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only