Thermo Scientific Chemicals

PIPES, 98.5+%, for biochemistry

Catalog number: 172611000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

PIPES, 98.5+%, for biochemistry

Catalog number: 172611000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 172611000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5625-37-6
IUPAC Name
sodium 2-[4-(2-sulfoethyl)piperazin-1-yl]ethane-1-sulfonate
Molecular Formula
C8H17N2NaO6S2
InChI Key
OGGAIRCLBMGXCZ-UHFFFAOYSA-M
SMILES
[Na+].OS(=O)(=O)CCN1CCN(CCS([O-])(=O)=O)CC1
Infrared spectrum
Conforms
Loss on drying
=<0.5 %
UV
(0.1 M in 0.1 M NaOH) 1 cm cell
UV
at 280 nm A: =<0.04
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text