Thermo Scientific Chemicals

CAPS, 99%, for biochemistry

Catalog number: 172620010
1 kg, Glass Bottle, Each
Thermo Scientific Chemicals

CAPS, 99%, for biochemistry

Catalog number: 172620010
1 kg, Glass Bottle, Each
Quantity
Packaging
Catalog number: 172620010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1135-40-6
IUPAC Name
3-(cyclohexylamino)propane-1-sulfonic acid
Molecular Formula
C9H19NO3S
InChI Key
PJWWRFATQTVXHA-UHFFFAOYSA-N
SMILES
OS(=O)(=O)CCCNC1CCCCC1
Infrared spectrum
Conforms
UV
(0.1 M in water)
Appearance (Color)
White
Titration with NaOH
>=98.5 % (On dry substance)
UV
at 260 nm A: =<0.04

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text